missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropylbenzolboronsäure, 97 %, Thermo Scientific Chemicals

Catalog Number p-7048024
Click to view available options
Menge:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
89787-12-2
C9H13BO2
164.01
MFCD03411937
KTZUVUWIBZMHMC-UHFFFAOYSA-N
2-isopropylphenylboronic acid, 2-isopropylphenyl boronic acid, 2-isopropylbenzeneboronic acid, 2-cumylboronic acid, 2-propan-2-ylphenyl boronic acid, o-isopropylphenylboronic acid, 2-isopropylphenboronic acid, 2-isopropylphenyboronic acid, 2-propan-2-yl phenyl boronic acid, boronic acid, 2-1-methylethyl phenyl
2773477
(2-propan-2-ylphenyl)boronsäure
CC(C)C1=CC=CC=C1B(O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
89787-12-2
C9H13BO2
164.01
MFCD03411937
KTZUVUWIBZMHMC-UHFFFAOYSA-N
2-isopropylphenylboronic acid, 2-isopropylphenyl boronic acid, 2-isopropylbenzeneboronic acid, 2-cumylboronic acid, 2-propan-2-ylphenyl boronic acid, o-isopropylphenylboronic acid, 2-isopropylphenboronic acid, 2-isopropylphenyboronic acid, 2-propan-2-yl phenyl boronic acid, boronic acid, 2-1-methylethyl phenyl
2773477
(2-propan-2-ylphenyl)boronsäure
CC(C)C1=CC=CC=C1B(O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Isopropylbenzeneboronic acid
Schmelzpunkt 85°C to 86°C
Menge 250 mg
Beilstein 3090298
Formelmasse 164.01
Reinheit (%) 97%

RUO – Nur für Forschungszwecke

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'