missing translation for 'onlineSavingsMsg'
Learn More

2-Mercapto-5-methyl-1,3,4-thiadiazol, 99 %, Thermo Scientific Chemicals

Catalog Number p-7025715
Click to view available options
Menge:
50 g
250 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
29490-19-5
C3H4N2S2
132.20
MFCD00038349
FPVUWZFFEGYCGB-UHFFFAOYSA-N
2-mercapto-5-methyl-1,3,4-thiadiazole, 5-methyl-1,3,4-thiadiazole-2-thiol, 1,3,4-thiadiazole-2 3h-thione, 5-methyl, 5-methyl-1,3,4-thiadiazole-2 3h-thione, 5-methyl-1,3,4-thiadiazol-2-thiol, 2-methyl-5-mercapto-1,3,4-thiadiazole, d2-1,3,4-thiadiazoline-5-thione, 2-methyl, cefazolin impurity e, pubchem9932, acmc-1cbwn
1810203
5-Methyl-3H-1,3,4-Thiadiazol-2-Thion
CC1=NNC(=S)S1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
29490-19-5
C3H4N2S2
132.20
MFCD00038349
FPVUWZFFEGYCGB-UHFFFAOYSA-N
2-mercapto-5-methyl-1,3,4-thiadiazole, 5-methyl-1,3,4-thiadiazole-2-thiol, 1,3,4-thiadiazole-2 3h-thione, 5-methyl, 5-methyl-1,3,4-thiadiazole-2 3h-thione, 5-methyl-1,3,4-thiadiazol-2-thiol, 2-methyl-5-mercapto-1,3,4-thiadiazole, d2-1,3,4-thiadiazoline-5-thione, 2-methyl, cefazolin impurity e, pubchem9932, acmc-1cbwn
1810203
5-Methyl-3H-1,3,4-Thiadiazol-2-Thion
CC1=NNC(=S)S1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Mercapto-5-methyl-1,3,4-thiadiazole
Schmelzpunkt 182°C to 185°C
Menge 50 g
Beilstein 606663
Formelmasse 132.21
Reinheit (%) 99%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'