missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethanol, +99 %, Thermo Scientific Chemicals

Catalog Number p-3758306
Click to view available options
Menge:
100 mL
1 L
109-86-4
C3H8O2
76.09
MFCD00002867
XNWFRZJHXBZDAG-UHFFFAOYSA-N
methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
8019
CHEBI:46790
2-Methoxyethanol
COCCO
109-86-4
C3H8O2
76.09
MFCD00002867
XNWFRZJHXBZDAG-UHFFFAOYSA-N
methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
8019
CHEBI:46790
2-Methoxyethanol
COCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Methoxyethanol
Namenshinweis Extra Dry
CAS Min. % 99.0
CAS Max. % 100.0
Schmelzpunkt -85.0°C
Farbe Farblos
Dichte 0.9600g/mL
Siedepunkt 124.0°C
Flammpunkt 38°C
Prozentgehaltsbereich 99% min. (GC)
Verpackung AcroSeal Glasflasche
Strukturformel CH3OCH2CH2OH
Brechungsindex 1.402
Menge 100 mL
Rückstand nach Verdampfung 0.001% max.
Merck Index 15, 6063
Relative Dichte 0.96
Löslichkeitsinformationen Solubility in water: soluble
Viskosität 1.7 mPa.s (20°C)
Formelmasse 76.09
Reinheit (%) 99+%
Güte Extra trocken
Physikalische Form Flüssig
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'