missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyphenyl isocyanate, 98%

Product Code. 11419226
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419226 25 g 25g
11409226 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419226 Supplier Thermo Scientific Alfa Aesar Supplier No. A18075.14
  • ONLINE EXCLUSIVE PRICE
    149.00 €
    134.65 € / 25 Gramm
    Sparen 14.35 €
    9% Rabatt
 Restricted Item
Lager vorrätig
Add to basket
This item is not returnable. View return policy

CAS: 700-87-8 | C8H7NO2 | 149.15 g/mol

2-Methoxyphenyl isocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxyphenyl isocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Hydrolyzes with water.

Notes
Moisture Sensitive. Protect from water and humidity. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 700-87-8
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.15
MDL Number MFCD00002004
InChI Key SUVCZZADQDCIEQ-UHFFFAOYSA-N
Synonym 2-methoxyphenyl isocyanate, 2-methoxyphenylisocyanate, benzene, 1-isocyanato-2-methoxy, o-methoxyphenyl isocyanate, 2-isocyanatoanisole, 1-isocyanato-2-methoxy-benzene, o-anisylisocyanate, 2-methoxybenzenisocyanate, o-anisyl isocyanate, methoxyphenyl isocyanate
PubChem CID 69695
IUPAC Name 1-isocyanato-2-methoxybenzene
SMILES COC1=CC=CC=C1N=C=O

Specifications

Density 1.122
Boiling Point 94°C to 96°C (10 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.5414
Quantity 25 g
UN Number UN2206
Beilstein 387700
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes with water.
Formula Weight 149.15
Percent Purity 98%
Chemical Name or Material 2-Methoxyphenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.