missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-[4-(methylsulfonyl)phenyl]-1-propanon, TRC

Artikelnummer. 30482580
Change view
Click to view available options
Menge:
10 mg
Unit Size:
10 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30482580 10 mg 10 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30482580 Supplier Toronto Research Chemicals Supplier No. TRCM32305010MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material 2-Methyl-1-[4-(methylsulfonyl)phenyl]-1-propanone
CAS 53207-59-3
Summenformel C11H14O3S
Menge 10 mg
Synonym p-Methylsulfonylisobutyrophenone, 2-Methyl-1-[4-(methylsulfonyl)phenyl]-1-propanone
InChI-Formel InChI=1S/C11H14O3S/c1-8(2)11(12)9-4-6-10(7-5-9)15(3,13)14/h4-8H,1-3H3
SMILES CC(C)C(=O)c1ccc(cc1)S(=O)(=O)C
IUPAC-Name 2-Methyl-1-(4-methylsulfonylphenyl)propan-1-on
Molekulargewicht (g/mol) 226.29
Formelmasse 226.0664
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.