missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-Thiopseudoharnstoff-Hämisulfat 98 %, Thermo Scientific Chemicals

Catalog Number p-168249
Click to view available options
Menge:
5 g
100 g
500 g
2.5 kg
Verpackung:
Glass Bottle
Kunststoffflasche
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
867-44-7
MFCD00013055
BZZXQZOBAUXLHZ-UHFFFAOYSA-N
s-methylisothiourea hemisulfate, 2-methyl-2-thiopseudourea hemisulfate, carbamimidothioic acid, methyl ester, sulfate 2:1, s-methylisothiourea sulfate 2:1, s-methylthiouronium sulfate 2:1, bis 2-methylisothiouronium sulphate, methyl carbamimidothioate sulfate 2:1, methylcarbamimidothioate sulfate 2:1, s-methylisothiourea, sulfate, 2-methyl-2-thiopseudourea sulfate 2:1
13347
methyl carbamimidothioate;sulfuric acid
CSC(=N)N.CSC(=N)N.OS(=O)(=O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
867-44-7
MFCD00013055
BZZXQZOBAUXLHZ-UHFFFAOYSA-N
s-methylisothiourea hemisulfate, 2-methyl-2-thiopseudourea hemisulfate, carbamimidothioic acid, methyl ester, sulfate 2:1, s-methylisothiourea sulfate 2:1, s-methylthiouronium sulfate 2:1, bis 2-methylisothiouronium sulphate, methyl carbamimidothioate sulfate 2:1, methylcarbamimidothioate sulfate 2:1, s-methylisothiourea, sulfate, 2-methyl-2-thiopseudourea sulfate 2:1
13347
methyl carbamimidothioate;sulfuric acid
CSC(=N)N.CSC(=N)N.OS(=O)(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Methyl-2-thiopseudourea hemisulfate
Namenshinweis 98%
CAS Min. % 97.5
CAS Max. % 100.0
Schmelzpunkt 240.0°C to 241.0°C
Farbe White
Infrarotspektrum Authentic
Prozentgehaltsbereich 98%
Verpackung Glass Bottle
Summenformel C4H14N4O4S3
Strukturformel [CH3SC(=NH)NH2]2·1/2H2SO4
Menge 5 g
Molekulargewicht (g/mol) 278.36
Formelmasse 139.18
Reinheit (%) 98%
Physikalische Form Crystalline Powder or Crystals
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'