missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-n-propyl-1,3-oxathian, cis + trans, 99 %, Thermo Scientific™

Catalog Number p-7026570
Click to view available options
Menge:
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
67715-80-4
C8H16OS
160.275
MFCD00673203
GKGOLPMYJJXRGD-UHFFFAOYSA-N
1,3-oxathiane, 2-methyl-4-propyl, 2-methyl-4-propyl-1,3-oxathiane, mixture of cis and trans, 1,3-oxathiane, 2-methyl-4-propyl-, 2r,4r-rel, 1,3-oxathiane, 2-methyl-4-propyl-, 2r,4s-rel, tropathiane, z-2-methyl-4-propyl-1,3-oxathiane, 2-tropical oxathiane, e-galbanum oxathiane, z-galbanum oxathiane, dsstox_cid_27704
101010
CHEBI:87335
2-Methyl-4-Propyl-1,3-Oxathian
CCCC1CCOC(S1)C
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
67715-80-4
C8H16OS
160.275
MFCD00673203
GKGOLPMYJJXRGD-UHFFFAOYSA-N
1,3-oxathiane, 2-methyl-4-propyl, 2-methyl-4-propyl-1,3-oxathiane, mixture of cis and trans, 1,3-oxathiane, 2-methyl-4-propyl-, 2r,4r-rel, 1,3-oxathiane, 2-methyl-4-propyl-, 2r,4s-rel, tropathiane, z-2-methyl-4-propyl-1,3-oxathiane, 2-tropical oxathiane, e-galbanum oxathiane, z-galbanum oxathiane, dsstox_cid_27704
101010
CHEBI:87335
2-Methyl-4-Propyl-1,3-Oxathian
CCCC1CCOC(S1)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Methyl-4-n-propyl-1,3-oxathiane
Namenshinweis cis + trans
Siedepunkt 218°C to 220°C
Menge 5 g
UN-Nummer UN1993
Formelmasse 160.27
Reinheit (%) 99%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'