Learn More
2-Methyl-4-(Trifluormethoxy)phenol, 97 %, Thermo Scientific™
Catalog Number p-7032480
Click to view available options
Menge:
250 mg
1 g
129676-67-1
C8H7F3O2
192.14
MFCD04972927
QMIXWHAIFDQEDH-UHFFFAOYSA-N
2-methyl-4-trifluoromethoxy phenol, 2-methyl-4-trifluoromethoxy-phenol
2783321
2-methyl-4-(trifluoromethoxy)phenol
CC1=C(O)C=CC(OC(F)(F)F)=C1
129676-67-1
C8H7F3O2
192.14
MFCD04972927
QMIXWHAIFDQEDH-UHFFFAOYSA-N
2-methyl-4-trifluoromethoxy phenol, 2-methyl-4-trifluoromethoxy-phenol
2783321
2-methyl-4-(trifluoromethoxy)phenol
CC1=C(O)C=CC(OC(F)(F)F)=C1
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Methyl-4-(trifluoromethoxy)phenol
Menge 250 mg
Formelmasse 192.14
Reinheit (%) 97%