missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-(-1-pyrrolidinylsulfonyl)benzenboronsäure, 98 %, Thermo Scientific™

Catalog Number 15410176
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
871333-01-6
C11H16BNO4S
269.122
MFCD08056365
IWOBOEZYVVBMGG-UHFFFAOYSA-N
2-methyl-5-pyrrolidin-1-ylsulfonyl phenyl boronic acid, 2-methyl-5-pyrrolidin-1-ylsulfonyl phenylboronic acid, 2-methyl-5-pyrrolidin-1-ylsulphonyl benzeneboronic acid, 2-methyl-5-pyrrolidine-1-sulfonyl phenylboronic acid, boronic acid,b-2-methyl-5-1-pyrrolidinylsulfonyl phenyl, acmc-209qhu, 2-borono-4-pyrrolidin-1-ylsulphonyl toluene, 1-3-borono-4-methylbenzene sulphonyl pyrrolidine, 2-methyl-5-pyrrolizinosulfonyl phenyl boronic acid, 2-methyl-5-pyrrolidin-1-ylsulfonyl phenyl boronicacid
44119827
(2-methyl-5-pyrrolidin-1-ylsulfonylphenyl)boronsäure
B(C1=C(C=CC(=C1)S(=O)(=O)N2CCCC2)C)(O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
871333-01-6
C11H16BNO4S
269.122
MFCD08056365
IWOBOEZYVVBMGG-UHFFFAOYSA-N
2-methyl-5-pyrrolidin-1-ylsulfonyl phenyl boronic acid, 2-methyl-5-pyrrolidin-1-ylsulfonyl phenylboronic acid, 2-methyl-5-pyrrolidin-1-ylsulphonyl benzeneboronic acid, 2-methyl-5-pyrrolidine-1-sulfonyl phenylboronic acid, boronic acid,b-2-methyl-5-1-pyrrolidinylsulfonyl phenyl, acmc-209qhu, 2-borono-4-pyrrolidin-1-ylsulphonyl toluene, 1-3-borono-4-methylbenzene sulphonyl pyrrolidine, 2-methyl-5-pyrrolizinosulfonyl phenyl boronic acid, 2-methyl-5-pyrrolidin-1-ylsulfonyl phenyl boronicacid
44119827
(2-methyl-5-pyrrolidin-1-ylsulfonylphenyl)boronsäure
B(C1=C(C=CC(=C1)S(=O)(=O)N2CCCC2)C)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Methyl-5-(-1-pyrrolidinylsulfonyl)benzeneboronic acid
Schmelzpunkt 126°C to 128°C
Menge 250 mg
Formelmasse 269.12
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'