missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-(pentafluorthio)benzylbromid, 96 %

Catalog Number p-7032461
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1261573-60-7
C9H8BrF3O
269.061
MFCD18399774
BSYHAIGVGUZQGK-UHFFFAOYSA-N
2-methyl-5-trifluoromethoxy benzyl bromide, 2-bromomethyl-1-methyl-4-trifluoromethoxy benzene
86277659
2-(brommethyl)-1-methyl-4-(trifluormethoxy)benzol
CC1=C(C=C(C=C1)OC(F)(F)F)CBr
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1261573-60-7
C9H8BrF3O
269.061
MFCD18399774
BSYHAIGVGUZQGK-UHFFFAOYSA-N
2-methyl-5-trifluoromethoxy benzyl bromide, 2-bromomethyl-1-methyl-4-trifluoromethoxy benzene
86277659
2-(brommethyl)-1-methyl-4-(trifluormethoxy)benzol
CC1=C(C=C(C=C1)OC(F)(F)F)CBr

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Methyl-5-(trifluoromethoxy)benzyl bromide
Menge 1 g
UN-Nummer UN3265
Empfindlichkeit Moisture sensitive
Formelmasse 269.06
Reinheit (%) 96%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'