missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)-oxazol-[5,4-c]-pyridin, Thermo Scientific™

Catalog Number p-7033840
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
169205-96-3
C7H6N2OS
166.20
MFCD08447111
DEGNWJBCNBPOPI-UHFFFAOYSA-N
2-methylthio oxazolo 5,4-c pyridine, 2-methylsulfanyl-oxazolo 5,4-c pyridine, acmc-1bzgj, 2-methylsulfanyloxazolo 5,4-c pyridine, 2-methylsulfanyl 1,3 oxazolo 5,4-c pyridine, 2-methylsulfanyl pyrido 4,3-d 1,3 oxazole, 2-methylsulfanyl-1,3 oxazolo 5,4-c pyridine
10241148
2-Methylsulfanyl-[1,3]Oxazolo[5,4-c]Pyridin
CSC1=NC2=CC=NC=C2O1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
169205-96-3
C7H6N2OS
166.20
MFCD08447111
DEGNWJBCNBPOPI-UHFFFAOYSA-N
2-methylthio oxazolo 5,4-c pyridine, 2-methylsulfanyl-oxazolo 5,4-c pyridine, acmc-1bzgj, 2-methylsulfanyloxazolo 5,4-c pyridine, 2-methylsulfanyl 1,3 oxazolo 5,4-c pyridine, 2-methylsulfanyl pyrido 4,3-d 1,3 oxazole, 2-methylsulfanyl-1,3 oxazolo 5,4-c pyridine
10241148
2-Methylsulfanyl-[1,3]Oxazolo[5,4-c]Pyridin
CSC1=NC2=CC=NC=C2O1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-(Methylthio)oxazolo[5,4-c]pyridine
Schmelzpunkt 79°C to 81°C
Menge 250 mg
Formelmasse 166.2
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'