missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Monodehydroxy-2'-chloro Ganciclovir, TRC
Spezifikation
Spezifikation
| Chemischer Name oder Material | 2'-Monodehydroxy-2'-chloro Ganciclovir |
| CAS | 108436-36-8 |
| Summenformel | C9 H12 Cl N5 O3 |
| Menge | 50 mg |
| Synonym | 2-Amino-9-[[(1RS)-2-chloro-1-(hydroxymethyl)ethoxy]methyl]-1,9-dihydro-6H-purin-6-one, Monochloroganciclovir, Ganciclovir Imp. C (EP), 6H-Purin-6-one, 2-amino-9-[[2-chloro-1-(hydroxymethyl)ethoxy]methyl]-1,9-dihydro-, 2-Amino-9-[[2-chloro-1-(hydroxymethyl)ethoxy]methyl]-1,9-dihydro-6H-purin-6-one, 2'-Monodehydroxy-2'-chloro Ganciclovir, Valganciclovir Hydrochloride Imp. I (EP) |
| InChI-Formel | InChI=1S/C9H12ClN5O3/c10-1-5(2-16)18-4-15-3-12-6-7(15)13-9(11)14-8(6)17/h3,5,16H,1-2,4H2,(H3,11,13,14,17) |
| SMILES | NC1=Nc2c(ncn2COC(CO)CCl)C(=O)N1 |
| IUPAC-Name | 2-Amino-9-[(1-chlor-3-hydroxypropan-2-yl)oxymethyl]-1H-purin-6-on |
| Molekulargewicht (g/mol) | 273.68 |
| Formelmasse | 273.0629 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?