missing translation for 'onlineSavingsMsg'
Learn More

2-(Morpholinoethyl)acrylamide, TRC

Artikelnummer. 30475087
Change view
Click to view available options
Menge:
100 mg
250 mg
500 mg
Unit Size:
100 Milligramm
250 Milligramm
500 Milligramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30475087 100 mg 100 Milligramm
30470170 250 mg 250 Milligramm
30470339 500 mg 500 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30475087 Supplier Toronto Research Chemicals Supplier No. TRCM732950100MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13276-17-0
Summenformel C9H16N2O2
Molekulargewicht (g/mol) 184.24
Synonym N-[2-(4-Morpholinyl)ethyl]-2-propenamide, N-(2-Morpholinoethyl)acrylamide, N-[2-(4-Morpholinyl)ethyl]acrylamide
IUPAC-Name N-(2-morpholin-4-ylethyl)prop-2-enamide
SMILES C=CC(=O)NCCN1CCOCC1

Specifications

Chemischer Name oder Material 2-(Morpholinoethyl)acrylamide
Menge 100 mg
InChI-Formel InChI=1S/C9H16N2O2/c1-2-9(12)10-3-4-11-5-7-13-8-6-11/h2H,1,3-8H2,(H,10,12)
Formelmasse 184.1212
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.