missing translation for 'onlineSavingsMsg'
Learn More

2-(n-Propylthio)nicotinamid, 97 %, Thermo Scientific™

Catalog Number p-7028120
Click to view available options
Menge:
0.25 g
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
175135-26-9
C9H12N2OS
196.27
MFCD00052904
JIYBJKGFAMWNDO-UHFFFAOYSA-N
2-propylthio nicotinamide, 2-n-propylthio nicotinamide, 2-propylsulfanyl pyridine-3-carboxamide, 2-propylthiopyridine-3-carboxamide, maybridge1_001275, 2-propylthio-3-pyridinecarboxamide, 3-pyridinecarboxamide,2-propylthio
2774535
2-(propylsulfanyl)pyridine-3-carboxamide
CCCSC1=NC=CC=C1C(N)=O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
175135-26-9
C9H12N2OS
196.27
MFCD00052904
JIYBJKGFAMWNDO-UHFFFAOYSA-N
2-propylthio nicotinamide, 2-n-propylthio nicotinamide, 2-propylsulfanyl pyridine-3-carboxamide, 2-propylthiopyridine-3-carboxamide, maybridge1_001275, 2-propylthio-3-pyridinecarboxamide, 3-pyridinecarboxamide,2-propylthio
2774535
2-(propylsulfanyl)pyridine-3-carboxamide
CCCSC1=NC=CC=C1C(N)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-(n-Propylthio)nicotinamide
Schmelzpunkt 146°C to 148°C
Menge 0.25 g
Formelmasse 196.27
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'