missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-4-Penten-2-ol, 98 %, Thermo Scientific™

Catalog Number 11429726
Click to view available options
Menge:
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
4743-74-2
C11H14O
162.232
MFCD00060886
BELRNEPYFJNSPN-UHFFFAOYSA-N
2-phenyl-4-penten-2-ol, 2-phenyl 4-penten-2-ol, allyl methyl phenyl carbinol, phenyl-methyl-allyl-carbinol
143761
2-phenylpent-4-en-2-ol
CC(CC=C)(C1=CC=CC=C1)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
4743-74-2
C11H14O
162.232
MFCD00060886
BELRNEPYFJNSPN-UHFFFAOYSA-N
2-phenyl-4-penten-2-ol, 2-phenyl 4-penten-2-ol, allyl methyl phenyl carbinol, phenyl-methyl-allyl-carbinol
143761
2-phenylpent-4-en-2-ol
CC(CC=C)(C1=CC=CC=C1)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Phenyl-4-penten-2-ol
Dichte 0.99
Menge 5 g
Formelmasse 162.23
Reinheit (%) 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'