Learn More
2-Phenylbuttersäure-d5-Methylester, TRC
Spezifikation
Specifications
| Chemischer Name oder Material | 2-Phenylbutyric Acid-d5 Methyl Ester |
| CAS | 1794940-35-4 |
| Summenformel | C11H9D5O2 |
| Menge | 50 mg |
| Synonym | 2-(Ethyl-d5)-2-phenylacetic Acid Methyl Ester, alpha-(Ethyl-d5)benzeneacetic Acid Methyl Ester, (RS)-2-Phenylbutanoic Acid-d5 Methyl Ester, (+/-)-2-Phenylbutanoic Acid-d5 Methyl Ester, alpha-(Ethyl-d5)-alpha-toluic Acid Methyl Ester, alpha-(Ethyl-d5)benzeneacetic Acid Methyl Ester, alpha-(Ethyl-d5)phenylacetic Acid Methyl Ester |
| InChI-Formel | InChI=1S/C11H14O2/c1-3-10(11(12)13-2)9-7-5-4-6-8-9/h4-8,10H,3H2,1-2H3/i1D3,3D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])C(C(=O)OC)c1ccccc1 |
| IUPAC-Name | Methyl-3,3,4,4,4-Pentadeuterio-2-phenylbutanoat |
| Molekulargewicht (g/mol) | 183.26 |
| Formelmasse | 183.1308 |