missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylindol-3-carboxaldehyd, 97 %, Thermo Scientific Chemicals

Catalog Number p-7025525
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
25365-71-3
C15H11NO
221.26
MFCD00435481
IFIFXODAHZPTEY-UHFFFAOYSA-N
2-phenylindole-3-carboxaldehyde, 2-phenylindole-3-carbaldehyde, 2-phenyl-3-formylindole, 3-formyl-2-phenylindole, 1h-indole-3-carboxaldehyde, 2-phenyl, 2-phenyl-3-formyl-indole, 3-formyl-2-phenyl indole, cambridge id 5402328, acmc-1cq37, 2-phenyl-3-indolecarboxaldehyde
613305
2-Phenyl-1H-Indol-3-Carbaldehyd
O=CC1=C(NC2=CC=CC=C12)C1=CC=CC=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
25365-71-3
C15H11NO
221.26
MFCD00435481
IFIFXODAHZPTEY-UHFFFAOYSA-N
2-phenylindole-3-carboxaldehyde, 2-phenylindole-3-carbaldehyde, 2-phenyl-3-formylindole, 3-formyl-2-phenylindole, 1h-indole-3-carboxaldehyde, 2-phenyl, 2-phenyl-3-formyl-indole, 3-formyl-2-phenyl indole, cambridge id 5402328, acmc-1cq37, 2-phenyl-3-indolecarboxaldehyde
613305
2-Phenyl-1H-Indol-3-Carbaldehyd
O=CC1=C(NC2=CC=CC=C12)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Phenylindole-3-carboxaldehyde
Schmelzpunkt 249°C to 253°C
Menge 1 g
Empfindlichkeit Air Sensitive
Formelmasse 221.26
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'