missing translation for 'onlineSavingsMsg'
Learn More

Azido-erythro-sphingosine, TRC

Artikelnummer. 30179193
Change view
Click to view available options
Menge:
1 mg
25 mg
250 mg
Packungsgröße:
1 Milligramm
25 Milligramm
250 Milligramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30179193 250 mg 250 Milligramm
30179605 25 mg 25 Milligramm
30179519 1 mg 1 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Product Code. 30179193 Supplier Toronto Research Chemicals Supplier No. TRCA926500250MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103348-49-8
Summenformel C18 H35 N3 O2
Molekulargewicht (g/mol) 325.49
IUPAC-Name (E,2S,3R)-2-azidooctadec-4-ene-1,3-diol
SMILES CCCCCCCCCCCCC\C=C\[C@@H](O)[C@H](CO)N=[N+]=[N-]

Specifications

Chemischer Name oder Material Azido-erythro-sphingosine
Menge 250 mg
InChI-Formel InChI=1S/C18H35N3O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(23)17(16-22)20-21-19/h14-15,17-18,22-23H,2-13,16H2,1H3/b15-14+/t17-,18+/m0/s1
Formelmasse 325.2729
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.