Learn More
(2S, 2'S)-6-Fluor-2-(2'-oxiranyl)chroman, TRC
Specifications
Specifications
| Chemischer Name oder Material | (2S, 2'S)-6-Fluoro-2-(2'-oxiranyl)chromane |
| CAS | 129050-23-3 |
| Summenformel | C11H11FO2 |
| Menge | 50 mg |
| Synonym | 2S)-2-((2S)-6-Fluoro-3,4-dihydro-2H-1-benzopyran-2-yl)oxirane, (2S)-6-Fluoro-3,4-dihydro-2-[(2S)-2-oxiranyl]-2H-1-Benzopyran |
| InChI-Formel | InChI=1S/C11H11FO2/c12-8-2-4-9-7(5-8)1-3-10(14-9)11-6-13-11/h2,4-5,10-11H,1,3,6H2/t10-,11-/m0/s1 |
| SMILES | Fc1ccc2O[C@@H](CCc2c1)[C@@H]3CO3 |
| IUPAC-Name | (2S)-6-Fluor-2-[(2S)-Oxiran-2-yl]-3,4-dihydro-2H-chromen |
| Molekulargewicht (g/mol) | 194.2 |
| Formelmasse | 194.0743 |