missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Piperazinyl)-1,2-benzisoxazol, 96 %

Catalog Number p-7033604
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
87691-89-2
C11H13N3O
203.245
MFCD08060968
ZDFQBFVFCPABKQ-UHFFFAOYSA-N
3-piperazin-1-yl benzo d isoxazole, 3-1-piperazinyl-1,2-benzisoxazole, 3-piperazin-1-yl-1,2-benzoxazole, 3-piperazin-1-yl-1,2-benzisoxazole, 3-1-piperazino-1,2-benzisoxazole, 3-piperazinylbenzo d isoxazole, 3-piperazin-1-yl-benzo d isoxazole, 3-1-piperazinyl-1,2-benzoxazole, 4 1,2-benzisoxazol-3-yl piperazine
13232475
3-Piperazin-1-yl-1,2-Benzoxazol
C1CN(CCN1)C2=NOC3=CC=CC=C32
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
87691-89-2
C11H13N3O
203.245
MFCD08060968
ZDFQBFVFCPABKQ-UHFFFAOYSA-N
3-piperazin-1-yl benzo d isoxazole, 3-1-piperazinyl-1,2-benzisoxazole, 3-piperazin-1-yl-1,2-benzoxazole, 3-piperazin-1-yl-1,2-benzisoxazole, 3-1-piperazino-1,2-benzisoxazole, 3-piperazinylbenzo d isoxazole, 3-piperazin-1-yl-benzo d isoxazole, 3-1-piperazinyl-1,2-benzoxazole, 4 1,2-benzisoxazol-3-yl piperazine
13232475
3-Piperazin-1-yl-1,2-Benzoxazol
C1CN(CCN1)C2=NOC3=CC=CC=C32

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-(1-Piperazinyl)-1,2-benzisoxazole
Schmelzpunkt 74°C to 76°C
Menge 250 mg
Formelmasse 203.25
Reinheit (%) 96%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'