missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Pyrrolidinylcarbonyl)-benzolboronsäurepinakolester, 97 %, Thermo Scientific™

Catalog Number p-7030453
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1073353-61-3
C17H24BNO3
301.19
MFCD09027086
CLPFNNNIZHMNPT-UHFFFAOYSA-N
pyrrolidin-1-yl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanone, 3-1-pyrrolidinylcarbonyl benzeneboronic acid pinacol ester, 3-pyrrolidine-1-carbonyl phenyl boronic acid pinacol ester, 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl pyrrolidine, amtb024, 3-pyrrolidinylcarbonylphenylboronic acid pinacol ester, 3-pyrrolidinylcarbonylphenylboronic acid, pinacol ester, 3-pyrrolidine-1-carbonyl benzeneboronic acid pinacol ester, 3-pyrrolidine-1-carbonyl phenylboronic acid pinacol ester, 3-pyrrolidine-1-carbonyl phenylboronic acid, pinacol ester
16218348
Pyrrolidin-1-yl-[3-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-yl)Phenyl]Methanon
CC1(C)OB(OC1(C)C)C1=CC(=CC=C1)C(=O)N1CCCC1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1073353-61-3
C17H24BNO3
301.19
MFCD09027086
CLPFNNNIZHMNPT-UHFFFAOYSA-N
pyrrolidin-1-yl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanone, 3-1-pyrrolidinylcarbonyl benzeneboronic acid pinacol ester, 3-pyrrolidine-1-carbonyl phenyl boronic acid pinacol ester, 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl pyrrolidine, amtb024, 3-pyrrolidinylcarbonylphenylboronic acid pinacol ester, 3-pyrrolidinylcarbonylphenylboronic acid, pinacol ester, 3-pyrrolidine-1-carbonyl benzeneboronic acid pinacol ester, 3-pyrrolidine-1-carbonyl phenylboronic acid pinacol ester, 3-pyrrolidine-1-carbonyl phenylboronic acid, pinacol ester
16218348
Pyrrolidin-1-yl-[3-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-yl)Phenyl]Methanon
CC1(C)OB(OC1(C)C)C1=CC(=CC=C1)C(=O)N1CCCC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-(1-Pyrrolidinylcarbonyl)benzeneboronic acid pinacol ester
Schmelzpunkt 96°C to 98°C
Menge 1 g
Formelmasse 301.19
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'