missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Pyrazol-3-yl)-pyridin, 95 %, Thermo Scientific™

Catalog Number p-7031927
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
45887-08-9
C8H7N3
145.165
MFCD03407951
JJLMOUMOJSUSSX-UHFFFAOYSA-N
3-1h-pyrazol-3-yl pyridine, 3-1h-pyrazol-5-yl pyridine, pyridine, 3-1h-pyrazol-3-yl, 3-3-pyridyl pyrazole, 3-pyridin-3-yl pyrazole, 3-3-pyridyl-1h-pyrazole, 5-3-pyridinyl-1h-pyrazole, 3-1h pyrazol-3-yl-pyridine, 3-1h-pyrazol-3-yl-pyridine, bestipharma 542-238
7074967
3-(1H-Pyrazol-5-yl)Pyridin
C1=CC(=CN=C1)C2=CC=NN2
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
45887-08-9
C8H7N3
145.165
MFCD03407951
JJLMOUMOJSUSSX-UHFFFAOYSA-N
3-1h-pyrazol-3-yl pyridine, 3-1h-pyrazol-5-yl pyridine, pyridine, 3-1h-pyrazol-3-yl, 3-3-pyridyl pyrazole, 3-pyridin-3-yl pyrazole, 3-3-pyridyl-1h-pyrazole, 5-3-pyridinyl-1h-pyrazole, 3-1h pyrazol-3-yl-pyridine, 3-1h-pyrazol-3-yl-pyridine, bestipharma 542-238
7074967
3-(1H-Pyrazol-5-yl)Pyridin
C1=CC(=CN=C1)C2=CC=NN2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-(1H-Pyrazol-3-yl)pyridine
Schmelzpunkt 57°C to 60°C
Menge 1 g
Formelmasse 145.16
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'