missing translation for 'onlineSavingsMsg'
Learn More

3-[2 -(Dimethylamino)ethylcarbamoyl]benzenboronsäure, 96 %, Thermo Scientific Chemicals

Catalog Number p-7034799
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
850567-31-6
C11H17BN2O3
236.078
MFCD04115706
WGZQGJKHTBTXLD-UHFFFAOYSA-N
3-2-dimethylamino ethylcarbamoyl phenylboronic acid, 3-2-dimethylamino ethyl carbamoyl phenyl boronic acid, 3-2-dimethylaminoethylaminocarbonyl benzeneboronic acid, 3-2-dimethylamino ethyl carbamoyl phenylboronic acid, acmc-209q0i, 3-2-dimethylamino ethyl carbamoyl-phenyl boronic acid, 3-2-dimethylamino ethyl carbamoyl phenyl boronicacid, 3-2-n,n-dimethylaminoethylaminocarbonyl benzene boronic acid
44119531
[3-[2-(Dimethylamino)ethylcarbamoyl]phenyl]boronsäure
B(C1=CC(=CC=C1)C(=O)NCCN(C)C)(O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
850567-31-6
C11H17BN2O3
236.078
MFCD04115706
WGZQGJKHTBTXLD-UHFFFAOYSA-N
3-2-dimethylamino ethylcarbamoyl phenylboronic acid, 3-2-dimethylamino ethyl carbamoyl phenyl boronic acid, 3-2-dimethylaminoethylaminocarbonyl benzeneboronic acid, 3-2-dimethylamino ethyl carbamoyl phenylboronic acid, acmc-209q0i, 3-2-dimethylamino ethyl carbamoyl-phenyl boronic acid, 3-2-dimethylamino ethyl carbamoyl phenyl boronicacid, 3-2-n,n-dimethylaminoethylaminocarbonyl benzene boronic acid
44119531
[3-[2-(Dimethylamino)ethylcarbamoyl]phenyl]boronsäure
B(C1=CC(=CC=C1)C(=O)NCCN(C)C)(O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-[2-(Dimethylamino)ethylcarbamoyl]benzeneboronic acid
Schmelzpunkt 42°C to 45°C
Menge 250 mg
Formelmasse 236.08
Reinheit (%) 96%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'