missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Fluorphenylsulfamoyl)benzenboronsäure, 97 %, Thermo Scientific™

Catalog Number p-7036195
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1449142-51-1
C12H11BFNO4S
295.091
MFCD20265270
CSHNEUKEFGIIOZ-UHFFFAOYSA-N
3-2-fluorophenylsulfamoyl benzeneboronic acid, 3-2-fluorophenyl sulfamoyl phenylboronic acid
73995813
[3-[(2-fluorphenyl)sulfamoyl]phenyl]boronsäure
B(C1=CC(=CC=C1)S(=O)(=O)NC2=CC=CC=C2F)(O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1449142-51-1
C12H11BFNO4S
295.091
MFCD20265270
CSHNEUKEFGIIOZ-UHFFFAOYSA-N
3-2-fluorophenylsulfamoyl benzeneboronic acid, 3-2-fluorophenyl sulfamoyl phenylboronic acid
73995813
[3-[(2-fluorphenyl)sulfamoyl]phenyl]boronsäure
B(C1=CC(=CC=C1)S(=O)(=O)NC2=CC=CC=C2F)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-(2-Fluorophenylsulfamoyl)benzeneboronic acid
Menge 250 mg
Formelmasse 295.09
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'