missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Methoxyethylaminocarbonyl)-benzolboronsäurepinakolester, 97 %, Thermo Scientific™

Catalog Number p-7030387
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1073353-64-6
C16H24BNO4
305.18
MFCD09266187
GNJHHGGRMLSKNG-UHFFFAOYSA-N
n-2-methoxyethyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 3-2-methoxyethylaminocarbonyl benzeneboronic acid pinacol ester, 3-2-methoxyethylamine-1-carbonyl phenyl boronic acid pinacol ester, amtb047, 3-2-methoxyethylcarbamoyl phenylboronic acid, pinacol ester, n-2-methoxyethyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide
46739043
N-(2-Methoxyethyl)-3-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-yl)Benzamid
COCCNC(=O)C1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1073353-64-6
C16H24BNO4
305.18
MFCD09266187
GNJHHGGRMLSKNG-UHFFFAOYSA-N
n-2-methoxyethyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 3-2-methoxyethylaminocarbonyl benzeneboronic acid pinacol ester, 3-2-methoxyethylamine-1-carbonyl phenyl boronic acid pinacol ester, amtb047, 3-2-methoxyethylcarbamoyl phenylboronic acid, pinacol ester, n-2-methoxyethyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide
46739043
N-(2-Methoxyethyl)-3-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-yl)Benzamid
COCCNC(=O)C1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-(2-Methoxyethylaminocarbonyl)benzeneboronic acid pinacol ester
Schmelzpunkt 93°C to 98°C
Menge 1 g
Formelmasse 305.18
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'