missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Pyridyl)propionsäure, 97%, Thermo Scientific Chemicals

Catalog Number p-7039386
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
15197-75-8
C8H9NO2
151.165
MFCD00995159
AZYKGQOIAGPVCK-UHFFFAOYSA-N
3-pyridin-2-yl propanoic acid, 3-2-pyridyl propionic acid, 3-pyridin-2-yl-propionic acid, 2-pyridinepropanoic acid, 3-2-pyridinyl propanoic acid, pyridine-2-propionic acid, 2-3-pyridyl propionic acid, 3-2-pyridyl propanoic acid, 3-2-pyridyl-propionic acid, 2-pyridinepropanoicacid
564292
3-(2-Pyridinyl)propansäure
C1=CC=NC(=C1)CCC(=O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
15197-75-8
C8H9NO2
151.165
MFCD00995159
AZYKGQOIAGPVCK-UHFFFAOYSA-N
3-pyridin-2-yl propanoic acid, 3-2-pyridyl propionic acid, 3-pyridin-2-yl-propionic acid, 2-pyridinepropanoic acid, 3-2-pyridinyl propanoic acid, pyridine-2-propionic acid, 2-3-pyridyl propionic acid, 3-2-pyridyl propanoic acid, 3-2-pyridyl-propionic acid, 2-pyridinepropanoicacid
564292
3-(2-Pyridinyl)propansäure
C1=CC=NC(=C1)CCC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-(2-Pyridyl)propionic acid
Menge 1 g
Formelmasse 151.16
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'