missing translation for 'onlineSavingsMsg'
Learn More

3-[2-(Trifluormethyl)phenyl]propionsäure, 97 %, Thermo Scientific™

Catalog Number p-7030971
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
94022-99-8
C10H9F3O2
218.175
MFCD06823965
YTDVNFDGHHHPEE-UHFFFAOYSA-N
3-2-trifluoromethyl phenyl propanoic acid, 3-2-trifluoromethyl phenyl propionic acid, 2-trifluoromethyl hydrocinnamic acid, 2-trifluoromethyl benzenepropanoic acid, benzenepropanoic acid, 2-trifluoromethyl, 3-o-alpha,alpha,alpha-trifluorotolyl propionic acid, acmc-20a31j, 3-2-trifluoromethyl phenyl p, 2-2-carboxyethyl benzotrifluoride
15284809
CHEBI:60707
3-[2-(Trifluormethyl)phenyl]propansäure
C1=CC=C(C(=C1)CCC(=O)O)C(F)(F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
94022-99-8
C10H9F3O2
218.175
MFCD06823965
YTDVNFDGHHHPEE-UHFFFAOYSA-N
3-2-trifluoromethyl phenyl propanoic acid, 3-2-trifluoromethyl phenyl propionic acid, 2-trifluoromethyl hydrocinnamic acid, 2-trifluoromethyl benzenepropanoic acid, benzenepropanoic acid, 2-trifluoromethyl, 3-o-alpha,alpha,alpha-trifluorotolyl propionic acid, acmc-20a31j, 3-2-trifluoromethyl phenyl p, 2-2-carboxyethyl benzotrifluoride
15284809
CHEBI:60707
3-[2-(Trifluormethyl)phenyl]propansäure
C1=CC=C(C(=C1)CCC(=O)O)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-[2-(Trifluoromethyl)phenyl]propionic acid
Schmelzpunkt 79°C to 86°C
Menge 1 g
Formelmasse 218.17
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'