missing translation for 'onlineSavingsMsg'
Learn More

3,3,3-Trifluor-2-(Trifluormethyl)propionsäure, 97 %, Thermo Scientific™

Catalog Number p-7035668
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
564-10-3
C4H2F6O2
196.048
MFCD00464165
RAEAYTICAPHWJW-UHFFFAOYSA-N
3,3,3-trifluoro-2-trifluoromethyl propanoic acid, 2-trifluoromethyl-3,3,3-trifluoropropionic acid, 3,3,3-trifluoro-2-trifluoromethyl propionic acid, propanoic acid,3,3,3-trifluoro-2-trifluoromethyl, hexafluoroisobutyric acid, acmc-1amsz, cf3 2chco2h, 3,3,3-trifluoro-2-trifluoromethylpropionic acid, 3,3,3-trifluoro-2-trifluoromethyl-propionic acid, 3,3,3-tris fluoranyl-2-trifluoromethyl propanoic acid
385606
3,3,3-Trifluor-2-(Trifluormethyl)propansäure
C(C(=O)O)(C(F)(F)F)C(F)(F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
564-10-3
C4H2F6O2
196.048
MFCD00464165
RAEAYTICAPHWJW-UHFFFAOYSA-N
3,3,3-trifluoro-2-trifluoromethyl propanoic acid, 2-trifluoromethyl-3,3,3-trifluoropropionic acid, 3,3,3-trifluoro-2-trifluoromethyl propionic acid, propanoic acid,3,3,3-trifluoro-2-trifluoromethyl, hexafluoroisobutyric acid, acmc-1amsz, cf3 2chco2h, 3,3,3-trifluoro-2-trifluoromethylpropionic acid, 3,3,3-trifluoro-2-trifluoromethyl-propionic acid, 3,3,3-tris fluoranyl-2-trifluoromethyl propanoic acid
385606
3,3,3-Trifluor-2-(Trifluormethyl)propansäure
C(C(=O)O)(C(F)(F)F)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3,3,3-Trifluoro-2-(trifluoromethyl)propionic acid
Schmelzpunkt 46°C to 53°C
Siedepunkt 125°C
Strukturformel (CF3)2CHCO2H
Menge 1 g
Formelmasse 196.05
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'