missing translation for 'onlineSavingsMsg'
Learn More

3,3,3-Trifluoro-2-(trifluoromethyl)propionyl chloride, TRC

Artikelnummer. 30449979
Change view
Click to view available options
Menge:
250 mg
500 mg
2.5 g
Unit Size:
2.50 Gramm
250 Milligramm
500 Milligramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30449979 250 mg 250 Milligramm
30451407 500 mg 500 Milligramm
30453882 2.5 g 2.50 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30449979 Supplier Toronto Research Chemicals Supplier No. TRCT900115250MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 382-19-4
Summenformel C4 H Cl F6 O
Molekulargewicht (g/mol) 214.49
IUPAC-Name 3,3,3-trifluoro-2-(trifluoromethyl)propanoyl chloride
SMILES FC(F)(F)C(C(=O)Cl)C(F)(F)F

Specifications

Chemischer Name oder Material 3,3,3-Trifluoro-2-(trifluoromethyl)propionyl chloride
Menge 250 mg
InChI-Formel InChI=1S/C4HClF6O/c5-2(12)1(3(6,7)8)4(9,10)11/h1H
Formelmasse 213.962
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.