Learn More
3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, sodium salt, BLD Pharm™
Spezifikation
Spezifikation
| CAS | 34487-61-1 |
| Molecular Formula | C19H13NaO5S |
| MDL Number | MFCD00066901 |
| Quantity | 25 g |
| SMILES | [O-]C1=CC=C(C2(C3=CC=C(O)C=C3)C4=CC=CC=C4S(O2)(=O)=O)C=C1.[Na+] |
| Molecular Weight (g/mol) | 376.36 |
| Chemical Name or Material | 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, sodium salt |