missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Bromphenyl)-1H-Pyrazol, 98 %, Thermo Scientific Chemicals

Catalog Number p-7033342
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
149739-65-1
C9H7BrN2
223.07
MFCD01940433
NVRXIZHZQPRBKL-UHFFFAOYSA-N
3-3-bromophenyl-1h-pyrazole, 5-3-bromophenyl-1h-pyrazole, 5-3-bromo-phenyl-1h-pyrazole, 3-3-bromophenyl pyrazole, 3-3-bromophenyl-2h-pyrazole, 1h-pyrazole, 3-3-bromophenyl, 1-bromo-3-1h-pyrazol-3-yl benzene, 5-3-bromophenyl pyrazole, acmc-1c2w8, ksc494i0r
2735613
5-(3-Bromphenyl)-1H-Pyrazol
BrC1=CC=CC(=C1)C1=CC=NN1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
149739-65-1
C9H7BrN2
223.07
MFCD01940433
NVRXIZHZQPRBKL-UHFFFAOYSA-N
3-3-bromophenyl-1h-pyrazole, 5-3-bromophenyl-1h-pyrazole, 5-3-bromo-phenyl-1h-pyrazole, 3-3-bromophenyl pyrazole, 3-3-bromophenyl-2h-pyrazole, 1h-pyrazole, 3-3-bromophenyl, 1-bromo-3-1h-pyrazol-3-yl benzene, 5-3-bromophenyl pyrazole, acmc-1c2w8, ksc494i0r
2735613
5-(3-Bromphenyl)-1H-Pyrazol
BrC1=CC=CC(=C1)C1=CC=NN1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-(3-Bromophenyl)-1H-pyrazole
Menge 1 g
Formelmasse 223.07
Reinheit (%) 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'