Learn More
3-[(3-Ethoxy-3-Oxopropyl)carbamoyl]benzolboronsäure, 97 %, Thermo Scientific™
Catalog Number 15429895
Click to view available options
Menge:
250 mg
1 g
850567-28-1
C12H16BNO5
265.072
MFCD04115701
COZCKUUEUXAVCK-UHFFFAOYSA-N
ethyl 3-3-boronobenzamido propanoate, 3-3-ethoxy-3-oxopropyl carbamoyl benzeneboronic acid, 3-3-ethoxy-3-oxopropyl carbamoyl phenyl boronic acid, 3-3-ethoxy-3-oxopropylcarbamoyl phenylboronic acid, 3-3-ethoxy-3-oxopropyl carbamoyl phenylboronic acid, acmc-209q0f, ethyl n-3-boronobenzoyl-beta-alaninate, ethyl 3-3-borono benzamidopropionate, ethyl 3-3-boronobenzoylamino propionate
44119409
[3-[(3-Ethoxy-3-oxopropyl)carbamoyl]phenyl]boronsäure
B(C1=CC(=CC=C1)C(=O)NCCC(=O)OCC)(O)O
850567-28-1
C12H16BNO5
265.072
MFCD04115701
COZCKUUEUXAVCK-UHFFFAOYSA-N
ethyl 3-3-boronobenzamido propanoate, 3-3-ethoxy-3-oxopropyl carbamoyl benzeneboronic acid, 3-3-ethoxy-3-oxopropyl carbamoyl phenyl boronic acid, 3-3-ethoxy-3-oxopropylcarbamoyl phenylboronic acid, 3-3-ethoxy-3-oxopropyl carbamoyl phenylboronic acid, acmc-209q0f, ethyl n-3-boronobenzoyl-beta-alaninate, ethyl 3-3-borono benzamidopropionate, ethyl 3-3-boronobenzoylamino propionate
44119409
[3-[(3-Ethoxy-3-oxopropyl)carbamoyl]phenyl]boronsäure
B(C1=CC(=CC=C1)C(=O)NCCC(=O)OCC)(O)O
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-[(3-Ethoxy-3-oxopropyl)carbamoyl]benzeneboronic acid
Schmelzpunkt 128°C to 132°C
Formelmasse 265.07
Reinheit (%) 97%