missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Fluor-4-Methylphenyl)propionsäure, 97 %, Thermo Scientific™

Catalog Number 15467995
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
881189-62-4
C10H11FO2
182.194
MFCD09025364
DYVQAEHNFXNAEH-UHFFFAOYSA-N
3-3-fluoro-4-methylphenyl propanoic acid, 3-3-fluoro-4-methylphenyl propionic acid, 3-3-fluoro-4-methyl-phenyl-propionic acid
46737500
3-(3-Fluor-4-methylphenyl)propansäure
CC1=C(C=C(C=C1)CCC(=O)O)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
881189-62-4
C10H11FO2
182.194
MFCD09025364
DYVQAEHNFXNAEH-UHFFFAOYSA-N
3-3-fluoro-4-methylphenyl propanoic acid, 3-3-fluoro-4-methylphenyl propionic acid, 3-3-fluoro-4-methyl-phenyl-propionic acid
46737500
3-(3-Fluor-4-methylphenyl)propansäure
CC1=C(C=C(C=C1)CCC(=O)O)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-(3-Fluoro-4-methylphenyl)propionic acid
Schmelzpunkt 95°C to 97°C
Menge 250 mg
Formelmasse 182.19
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'