missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Methyl-2-pyridyl)-5-phenyl-1,2,4-oxadiazol, 97 %, Thermo Scientific™

Catalog Number p-7036391
Click to view available options
Menge:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1400191-51-6
C14H11N3O
237.26
MFCD20265320
ZHKWMFUDVLEAHG-UHFFFAOYSA-N
3-3-methyl-2-pyridyl-5-phenyl-1,2,4-oxadiazole, 3-methyl-2-5-phenyl-1,2,4-oxadiazol-3-yl pyridine, 3-3-methylpyridin-2-yl-5-phenyl-1,2,4-oxadiazole
73995920
3-(3-Methylpyridin-2-yl)-5-Phenyl-1,2,4-Oxadiazol
CC1=C(N=CC=C1)C1=NOC(=N1)C1=CC=CC=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1400191-51-6
C14H11N3O
237.26
MFCD20265320
ZHKWMFUDVLEAHG-UHFFFAOYSA-N
3-3-methyl-2-pyridyl-5-phenyl-1,2,4-oxadiazole, 3-methyl-2-5-phenyl-1,2,4-oxadiazol-3-yl pyridine, 3-3-methylpyridin-2-yl-5-phenyl-1,2,4-oxadiazole
73995920
3-(3-Methylpyridin-2-yl)-5-Phenyl-1,2,4-Oxadiazol
CC1=C(N=CC=C1)C1=NOC(=N1)C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-(3-Methyl-2-pyridyl)-5-phenyl-1,2,4-oxadiazole
Menge 250 mg
Formelmasse 237.26
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'