missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trifluorbenzophenon, 97 %, Thermo Scientific™

Catalog Number p-7029280
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
70028-88-5
C13H7F3O
MFCD00083525
3,4,5-trifluorobenzophenone, phenyl 3,4,5-trifluorophenyl methanone, phenyl-3,4,5-trifluorophenyl methanone, methanone,phenyl 3,4,5-trifluorophenyl, methanone, phenyl 3,4,5-trifluorophenyl, phenyl 3,4,5-trifluorophenyl ketone, phenyl-3,4,5-tris fluoranyl phenyl methanone
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
70028-88-5
C13H7F3O
MFCD00083525
3,4,5-trifluorobenzophenone, phenyl 3,4,5-trifluorophenyl methanone, phenyl-3,4,5-trifluorophenyl methanone, methanone,phenyl 3,4,5-trifluorophenyl, methanone, phenyl 3,4,5-trifluorophenyl, phenyl 3,4,5-trifluorophenyl ketone, phenyl-3,4,5-tris fluoranyl phenyl methanone

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3,4,5-Trifluorobenzophenone
Menge 1 g
Formelmasse 236.19
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'