Learn More
3,4-Dichlorobenzyl Chloride, TRC
Product Code. 30006017
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006017 25 g 25g
30006877 10 g 10g
30005254 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30006017 Supplier Toronto Research Chemicals Supplier No. TRCD43841525G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 102-47-6
Molecular Formula C7 H5 Cl3
Molecular Weight (g/mol) 195.47
Synonym alpha,3,4-Trichlorotoluene, Benzene, 1,2-dichloro-4-(chloromethyl)-, Toluene, α,3,4-trichloro- (7CI,8CI), 1,2-Dichloro-4-(chloromethyl)benzene, 3,4,α-Trichlorotoluene, 3,4-Dichloro-1-(chloromethyl)benzene, 3,4-Dichlorobenzyl chloride, 3,4-Dichlorophenylmethyl chloride, 4-Chloromethyl-1,2-dichlorobenzene, NSC 406893, α,3,4-Trichlorotoluene
IUPAC Name 1,2-dichloro-4-(chloromethyl)benzene
SMILES ClCc1ccc(Cl)c(Cl)c1
Specifications
Quantity 25 g
InChI Formula InChI=1S/C7H5Cl3/c8-4-5-1-2-6(9)7(10)3-5/h1-3H,4H2
Formula Weight 193.9457
Chemical Name or Material alpha,3,4-Trichlorotoluene