missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorsalicylsäure, 98 %, Thermo Scientific Chemicals

Catalog Number p-7028684
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
189283-51-0
C7H4F2O3
174.103
MFCD03428564
GWOOBUWKTOCYKY-UHFFFAOYSA-N
3,4-difluorosalicylic acid, benzoic acid, 3,4-difluoro-2-hydroxy, 3,4-difluoro-2-hydroxy-benzoic acid, acmc-20anf8, 3,4-difluoro-hydroxybenzoic acid, 3,4-difluro-2-hydroxybenzoic acid, 3,4-difluoro-2-hydroxy benzoic acid, 3,4-bis fluoranyl-2-oxidanyl-benzoic acid
2758286
3,4-difluor-2-hydroxybenzoesäure
C1=CC(=C(C(=C1C(=O)O)O)F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
189283-51-0
C7H4F2O3
174.103
MFCD03428564
GWOOBUWKTOCYKY-UHFFFAOYSA-N
3,4-difluorosalicylic acid, benzoic acid, 3,4-difluoro-2-hydroxy, 3,4-difluoro-2-hydroxy-benzoic acid, acmc-20anf8, 3,4-difluoro-hydroxybenzoic acid, 3,4-difluro-2-hydroxybenzoic acid, 3,4-difluoro-2-hydroxy benzoic acid, 3,4-bis fluoranyl-2-oxidanyl-benzoic acid
2758286
3,4-difluor-2-hydroxybenzoesäure
C1=CC(=C(C(=C1C(=O)O)O)F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3,4-Difluorosalicylic acid
Schmelzpunkt 177°C to 179°C
Menge 250 mg
Formelmasse 174.1
Reinheit (%) 98%

RUO – Nur für Forschungszwecke

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'