Learn More
3,4-Dihydro-2H-pyran-2-methanol, 96%
Product Code. 15472558
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15472558 5 g 5g
15462558 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15472558 Supplier Thermo Scientific Alfa Aesar Supplier No. H56124.06
 Restricted Item
Lager vorrätig
Add to basket
missing translation for 'validMainframeMsg'
CAS: 3749-36-8 | C6H10O2 | 114.144 g/mol
It is used in the preparation of supported dihydropyran for mild immobilization and cleavage of alcohols
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the preparation of supported dihydropyran for mild immobilization and cleavage of alcohols
Notes
Incompatible with oxidizing agents, acids and heats. Store at room temperature.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3749-36-8
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00051507
InChI Key XMICBFRKICBBKD-UHFFFAOYSA-N
Synonym 3,4-dihydro-2h-pyran-2-methanol, 2-hydroxymethyl-3,4-dihydro-2h-pyran, dhp linker, 3,4-dihydro-2h-pyran-2-yl methanol, 2h-pyran-2-methanol, 3,4-dihydro, dhp resin, ellmans dhp resin, zlchem 376, pubchem9703, acmc-209itf
PubChem CID 95559
IUPAC Name 3,4-dihydro-2H-pyran-2-ylmethanol
SMILES C1CC(OC=C1)CO
Specifications
Density 1.101
Boiling Point 92°C to 93°C (25 mmHg)
Flash Point 89°C (192°F)
Refractive Index 1.478
Quantity 5 g
Formula Weight 114.14
Percent Purity 96%
Chemical Name or Material 3,4-Dihydro-2H-pyran-2-methanol