missing translation for 'onlineSavingsMsg'
Learn More

3-[4-Fluor-3-(Trifluormethoxy)phenyl]propionsäure, 97 %, Thermo Scientific™

Catalog Number p-7032537
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1240256-80-7
C10H8F4O3
252.165
MFCD09999449
RDAQINCBAFDESL-UHFFFAOYSA-N
3-4-fluoro-3-trifluoromethoxy phenyl propionic acid, 3-4-fluoro-3-trifluoromethoxy phenyl propanoic acid, 3-4-fluoro-3-trifluoromethoxy phenyl propanoic a
66523471
3-[4-Fluor-3-(trifluormethoxy)phenyl]propansäure
C1=CC(=C(C=C1CCC(=O)O)OC(F)(F)F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1240256-80-7
C10H8F4O3
252.165
MFCD09999449
RDAQINCBAFDESL-UHFFFAOYSA-N
3-4-fluoro-3-trifluoromethoxy phenyl propionic acid, 3-4-fluoro-3-trifluoromethoxy phenyl propanoic acid, 3-4-fluoro-3-trifluoromethoxy phenyl propanoic a
66523471
3-[4-Fluor-3-(trifluormethoxy)phenyl]propansäure
C1=CC(=C(C=C1CCC(=O)O)OC(F)(F)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-[4-Fluoro-3-(trifluoromethoxy)phenyl]propionic acid
Menge 250 mg
Formelmasse 252.16
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'