Learn More
3,5-Bis(trifluoromethyl)benzonitrile, 98%, BLD Pharm™
Product Code. 30947424
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
1 kg
Unit Size:
100g
10g
1g
25g
500g
5g
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30947424 500 g 500g
30930214 1 kg -
30929882 100 g 100g
30932138 25 g 25g
30934152 10 g 10g
30930018 5 g 5g
30935154 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 30947424 Supplier BLD Pharm Supplier No. BD11443500g
 Restricted Item
Verfügbar ab: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
CAS 27126-93-8
Molecular Formula C9H3F6N
MDL Number MFCD00000379
Quantity 500 g
SMILES N#CC1=CC(C(F)(F)F)=CC(C(F)(F)F)=C1
Molecular Weight (g/mol) 239.12
Percent Purity 98%
Chemical Name or Material 3,5-Bis(trifluoromethyl)benzonitrile