missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluormethyl)benzophenon, 95 %, Thermo Scientific™

Catalog Number p-7024821
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
21221-93-2
C15H8F6O
318.218
MFCD00015510
QETSMUIBVUASOJ-UHFFFAOYSA-N
3,5-bis trifluoromethyl benzophenone, 3,5-bis trifluoromethyl phenyl phenyl methanone, fxffr cvr & exfff, acmc-1cotv, 3,5-di trifluoromethyl benzophenone, 3',5'-bis trifluoromethyl benzophenone, 3,5-bis trifluoromethyl phenyl phenyl ketone, 3,5-bis trifluoromethyl phenyl-phenylmethanone
88830
[3,5-bis(trifluormethyl)phenyl]-phenylmethanon
C1=CC=C(C=C1)C(=O)C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
21221-93-2
C15H8F6O
318.218
MFCD00015510
QETSMUIBVUASOJ-UHFFFAOYSA-N
3,5-bis trifluoromethyl benzophenone, 3,5-bis trifluoromethyl phenyl phenyl methanone, fxffr cvr & exfff, acmc-1cotv, 3,5-di trifluoromethyl benzophenone, 3',5'-bis trifluoromethyl benzophenone, 3,5-bis trifluoromethyl phenyl phenyl ketone, 3,5-bis trifluoromethyl phenyl-phenylmethanone
88830
[3,5-bis(trifluormethyl)phenyl]-phenylmethanon
C1=CC=C(C=C1)C(=O)C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3,5-Bis(trifluoromethyl)benzophenone
Siedepunkt 111°C to 112°C (0.2 mmHg)
Brechungsindex 1.496
Menge 1 g
Beilstein 3066566
Formelmasse 318.22
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'