missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di(trifluoromethyl)phenyl isothiocyanate, ≥97%, Thermo Scientific™

Product Code. 10107293
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
Product Code. Quantity unitSize
10107293 25 g 25g
10332923 10 g 10g
2 options
This item is not returnable. View return policy

Product Code. 10107293

Brand: Thermo Scientific Maybridge SB01673EB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 23165-29-9
Molecular Formula C9H3F6NS
Molecular Weight (g/mol) 271.18
MDL Number MFCD00040838
InChI Key FXOSSGVJGGNASE-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-3,5-bis trifluoromethyl benzene, 3,5-bis trifluoromethyl phenylisothiocyanate, isothiocyanic acid 3,5-bis trifluoromethyl phenyl ester, 3,5-di trifluoromethyl phenyl isothiocyanate, 3,5-bis-trifluoromethyl phenyl isothiocyanate, 3,5-bis-trifluoromethylphenyl isothiocyanate, benzene, 1-isothiocyanato-3,5-bis trifluoromethyl, pubchem5019, acmc-209g1w
PubChem CID 2733395
IUPAC Name 1-isothiocyanato-3,5-bis(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC(=CC(=C1)N=C=S)C(F)(F)F

Specifications

CAS Min % 97.0
Density 1.4850g/mL
Assay Percent Range 97+%
Packaging Amber glass bottle
Linear Formula (CF3)2C6H3NCS
Quantity 25 g
Formula Weight 271.18
Percent Purity 97%
Chemical Name or Material 3, 5-Di(trifluoromethyl)phenyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.