missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorbenzhydrol, 95 %, Thermo Scientific™

Catalog Number p-7030946
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
153877-56-6
C13H10F2O
220.219
MFCD06201256
MZBBRGLZQRMJPL-UHFFFAOYSA-N
3,5-difluorobenzhydrol, 3,5-difluorophenyl-phenylmethanol, 3,5-difluorophenyl phenyl methanol
2758253
(3,5-difluorphenyl)-Phenylmethanol
C1=CC=C(C=C1)C(C2=CC(=CC(=C2)F)F)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
153877-56-6
C13H10F2O
220.219
MFCD06201256
MZBBRGLZQRMJPL-UHFFFAOYSA-N
3,5-difluorobenzhydrol, 3,5-difluorophenyl-phenylmethanol, 3,5-difluorophenyl phenyl methanol
2758253
(3,5-difluorphenyl)-Phenylmethanol
C1=CC=C(C=C1)C(C2=CC(=CC(=C2)F)F)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3,5-Difluorobenzhydrol
Menge 1 g
Formelmasse 220.22
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'