missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorphenylessigsäure, 99 %, Thermo Scientific™

Catalog Number p-7024573
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
105184-38-1
C8H6F2O2
172.131
MFCD00010316
IGGNSAVLXJKCNH-UHFFFAOYSA-N
3,5-difluorophenylacetic acid, 2-3,5-difluorophenyl acetic acid, 3,5-difluorophenylaceticacid, 3,5-difluorobenzeneacetic acid, 3,5-difluorophenyl acetic acid, 3,5-difluoro-phenyl-acetic acid, benzeneacetic acid, 3,5-difluoro, 3,5-diifluorophenylacetic acid, 3,5-difluorophenylac
145424
2-(3,5-dichlorphenyl)Essigsäure
C1=C(C=C(C=C1F)F)CC(=O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
105184-38-1
C8H6F2O2
172.131
MFCD00010316
IGGNSAVLXJKCNH-UHFFFAOYSA-N
3,5-difluorophenylacetic acid, 2-3,5-difluorophenyl acetic acid, 3,5-difluorophenylaceticacid, 3,5-difluorobenzeneacetic acid, 3,5-difluorophenyl acetic acid, 3,5-difluoro-phenyl-acetic acid, benzeneacetic acid, 3,5-difluoro, 3,5-diifluorophenylacetic acid, 3,5-difluorophenylac
145424
2-(3,5-dichlorphenyl)Essigsäure
C1=C(C=C(C=C1F)F)CC(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3,5-Difluorophenylacetic acid
Schmelzpunkt 68°C to 70°C
Menge 1 g
Beilstein 3605064
Formelmasse 172.13
Reinheit (%) 99%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'