missing translation for 'onlineSavingsMsg'
Få mere at vide

3,5-Diiodthyrolaldehydmethylether, TRC

Artikelnummer. 30370915
Skift visning
Klik for at se tilgængelige muligheder
Menge:
250 mg
Pakningsstørrelse:
250 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30370915 250 mg 250 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30370915 Leverandør Toronto Research Chemicals Leverandørnr. TRCD455135250MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Denne vare kan ikke returneres. Se returpolicy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material 3,5-Diiodo Thyroaldehyde Methyl Ether
CAS 69240-57-9
Summenformel C14H10I2O3
Menge 250 mg
Synonym 3,5-Diiodo-4-(4-methoxyphenoxy)-benzaldehyde, 3,5-Diiodo-4-(p-methoxyphenoxy)-benzaldehyde
InChI-Formel InChI=1S/C14H10I2O3/c1-18-10-2-4-11(5-3-10)19-14-12(15)6-9(8-17)7-13(14)16/h2-8H,1H3
SMILES COc1ccc(Oc2c(I)cc(C=O)cc2I)cc1
IUPAC-Name 3,5-Diiod-4-(4-methoxyphenoxy)benzaldehyd
Molekulargewicht (g/mol) 480.04
Formelmasse 479.8719
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.