Learn More
3,6-Dichlor-2-fluorbenzylbromid, 97 %, Thermo Scientific™
Catalog Number p-7031752
Click to view available options
Menge:
1 g
5 g
916420-69-4
C7H4BrCl2F
257.91
MFCD09025389
RXWLZGRXMKRFEH-UHFFFAOYSA-N
3,6-dichloro-2-fluorobenzyl bromide, 2-bromomethyl-1,4-dichloro-3-fluorobenzene
46737555
2-(bromomethyl)-1,4-dichloro-3-fluorobenzene
FC1=C(Cl)C=CC(Cl)=C1CBr
916420-69-4
C7H4BrCl2F
257.91
MFCD09025389
RXWLZGRXMKRFEH-UHFFFAOYSA-N
3,6-dichloro-2-fluorobenzyl bromide, 2-bromomethyl-1,4-dichloro-3-fluorobenzene
46737555
2-(bromomethyl)-1,4-dichloro-3-fluorobenzene
FC1=C(Cl)C=CC(Cl)=C1CBr
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3,6-Dichloro-2-fluorobenzyl bromide
Schmelzpunkt 42°C to 45°C
Menge 1 g
UN-Nummer UN3261
Formelmasse 257.92
Reinheit (%) 97%