missing translation for 'onlineSavingsMsg'
Learn More

3-(6-Oxo-1,6-dihydro-3-pyridazinyl)-benzonitril, 97 %, Thermo Scientific™

Catalog Number 15461456
Click to view available options
Menge:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
52240-08-1
C11H7N3O
197.197
MFCD16482247
INFIYPRKBSENAH-UHFFFAOYSA-N
3-6-oxo-1,6-dihydropyridazin-3-yl benzonitrile, 3-6-oxo-1,6-dihydro-3-pyridazinyl benzonitrile, 3-6-oxo-1,6-dihydro-pyridazin-3-yl-benzonitrile, 3-6-oxo-1h-pyridazin-3-yl benzonitrile, 3-6-oxo-1,6-dihydropyridazin-3-yl-benzonitrile, benzonitrile, 3-1,6-dihydro-6-oxo-3-pyridazinyl
20326520
3-(6-Oxo-1H-Pyridazin-3-yl)Benzonitril
C1=CC(=CC(=C1)C2=NNC(=O)C=C2)C#N
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
52240-08-1
C11H7N3O
197.197
MFCD16482247
INFIYPRKBSENAH-UHFFFAOYSA-N
3-6-oxo-1,6-dihydropyridazin-3-yl benzonitrile, 3-6-oxo-1,6-dihydro-3-pyridazinyl benzonitrile, 3-6-oxo-1,6-dihydro-pyridazin-3-yl-benzonitrile, 3-6-oxo-1h-pyridazin-3-yl benzonitrile, 3-6-oxo-1,6-dihydropyridazin-3-yl-benzonitrile, benzonitrile, 3-1,6-dihydro-6-oxo-3-pyridazinyl
20326520
3-(6-Oxo-1H-Pyridazin-3-yl)Benzonitril
C1=CC(=CC(=C1)C2=NNC(=O)C=C2)C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-(6-Oxo-1,6-dihydro-3-pyridazinyl)benzonitrile
Menge 1 g
Formelmasse 197.19
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'