missing translation for 'onlineSavingsMsg'
Learn More

3-Acetyl-2,5-dichlorothiophene, 98%, ACROS Organics™

Product Code. 12742744 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12742744 5 g 5g
10381284 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12742744 Supplier Thermo Scientific Acros Supplier No. 309620050
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 36157-40-1
Molecular Formula C6H4Cl2OS
Molecular Weight (g/mol) 195.057
MDL Number MFCD00014522
InChI Key GYFDNIRENHZKGR-UHFFFAOYSA-N
Synonym 3-acetyl-2,5-dichlorothiophene, 1-2,5-dichlorothiophen-3-yl ethanone, 1-2,5-dichloro-3-thienyl ethan-1-one, 1-2,5-dichlorothiophen-3-yl ethan-1-one, 1-2,5-dichloro-3-thienyl ethanone, 2,5-dichloro-3-thienyl methyl ketone, ethanone, 1-2,5-dichloro-3-thienyl, 1-2,5-dichloro-thiophen-3-yl-ethanone, 1-2,5-dichlorothien-3-yl ethan-1-one, zlchem 481
PubChem CID 118920
IUPAC Name 1-(2,5-dichlorothiophen-3-yl)ethanone
SMILES CC(=O)C1=C(SC(=C1)Cl)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 36°C to 40°C
Color Brown to Tan
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 5 g
Formula Weight 195.07
Percent Purity ≥97.5%
Physical Form Chunks
Chemical Name or Material 3-Acetyl-2, 5-dichlorothiophene, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.