missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-Methylphenol, 98 %, Thermo Scientific Chemicals

Catalog Number p-7057159
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
53222-92-7
C7H9NO
123.155
MFCD00007787
FLROJJGKUKLCAE-UHFFFAOYSA-N
3-amino-o-cresol, phenol, 3-amino-2-methyl, 2-amino-o-cresol, 2-methyl-3-aminophenol, 3-hydroxy-2-methylaniline, aminokresol, o-cresol, 3-amino, pubchem15039, 2-methyl-3-amino-phenol, 3-amino-2-methyl phenol
104448
3-amino-2-methylphenol
CC1=C(C=CC=C1O)N
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
53222-92-7
C7H9NO
123.155
MFCD00007787
FLROJJGKUKLCAE-UHFFFAOYSA-N
3-amino-o-cresol, phenol, 3-amino-2-methyl, 2-amino-o-cresol, 2-methyl-3-aminophenol, 3-hydroxy-2-methylaniline, aminokresol, o-cresol, 3-amino, pubchem15039, 2-methyl-3-amino-phenol, 3-amino-2-methyl phenol
104448
3-amino-2-methylphenol
CC1=C(C=CC=C1O)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Amino-2-methylphenol
Schmelzpunkt 129°C to 130°C
Menge 1 g
Beilstein 2716070
Formelmasse 123.16
Reinheit (%) 98%
Physikalische Form Fest

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'