missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-Methoxybenzanilid, 98 %, Thermo Scientific™

Catalog Number p-7024463
Click to view available options
Menge:
10 g
50 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
120-35-4
C14H14N2O2
242.278
MFCD00017166
LHMQDVIHBXWNII-UHFFFAOYSA-N
3-amino-4-methoxybenzanilide, 3-amino-p-anisanilide, benzamide, 3-amino-4-methoxy-n-phenyl, 3-amino-4-methoxy benzanilide, benzanilide, 3-amino-4-methoxy, p-anisanilide, 3-amino, unii-v5y646aw8s, amba, 3-amino-4-methoxy-n-phenyl-benzamide, 3-amino-4-methoxyphenyl-n-benzamide
8426
3-amino-4-methoxy-N-phenylbenzamid
COC1=C(C=C(C=C1)C(=O)NC2=CC=CC=C2)N
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
120-35-4
C14H14N2O2
242.278
MFCD00017166
LHMQDVIHBXWNII-UHFFFAOYSA-N
3-amino-4-methoxybenzanilide, 3-amino-p-anisanilide, benzamide, 3-amino-4-methoxy-n-phenyl, 3-amino-4-methoxy benzanilide, benzanilide, 3-amino-4-methoxy, p-anisanilide, 3-amino, unii-v5y646aw8s, amba, 3-amino-4-methoxy-n-phenyl-benzamide, 3-amino-4-methoxyphenyl-n-benzamide
8426
3-amino-4-methoxy-N-phenylbenzamid
COC1=C(C=C(C=C1)C(=O)NC2=CC=CC=C2)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Amino-4-methoxybenzanilide
Schmelzpunkt 154°C to 156°C
Menge 10 g
Formelmasse 242.28
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'